• USA Home
  • 161462 - N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride (EDC) 98%

EMAIL THIS PAGE TO A FRIEND
161462 Aldrich

N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride (EDC) 98%

≥98%

Purchase

Properties

InChI Key   FPQQSJJWHUJYPU-UHFFFAOYSA-N
assay   ≥98%
mp   111-113 °C(lit.)

Description

Application

Coupling reagent used to form amide bonds in oligonucleotides and tetraazamacrocycles.

Water soluble carbodiimide for the preparation of chloroanthraquinone hydrazides, which unexpectedly cyclize to anthrapyrazoles (en route to developing histochemical probes).

Other Notes

This product has been replaced by E7750-Sigma-Aldrich | N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride commercial grade, powder

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
FF2200000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

E7750 N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, commercial grade, powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?