EMAIL THIS PAGE TO A FRIEND
166049 Aldrich

4-Benzyloxyaniline

  • CAS Number 6373-46-2

  • Empirical Formula (Hill Notation) C13H13NO

  • Molecular Weight 199.25

  •  PubChem Substance ID 329751280

Purchase

Properties

InChI Key   FIIDVVUUWRJXLF-UHFFFAOYSA-N

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?