• USA Home
  • 167517 - Phenyl chloroformate

EMAIL THIS PAGE TO A FRIEND
167517 Aldrich

Phenyl chloroformate

Synonym: Chloroformic acid phenyl ester

Purchase

Properties

vapor density   1 (vs air)
vapor pressure   1.22 psi ( 20 °C)
  1.67 psi ( 55 °C)
InChI Key   AHWALFGBDFAJAI-UHFFFAOYSA-N
refractive index   n20/D 1.511(lit.)
bp   74-75 °C/13 mmHg(lit.)
density   1.248 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2746 8(6.1) / PGII
WGK Germany 
3
RTECS 
FG3850000
Flash Point(F) 
156.2 °F
Flash Point(C) 
69 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?