EMAIL THIS PAGE TO A FRIEND
169471 Aldrich

Potassium nitrate

99%

Purchase

Properties

assay   99%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS03  GHS03
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1486 5.1 / PGIII
WGK Germany 
1
RTECS 
TT3700000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?