• USA Home
  • 176311 - Benzenediazonium hexafluorophosphate

EMAIL THIS PAGE TO A FRIEND
176311 Aldrich

Benzenediazonium hexafluorophosphate

  • CAS Number 369-58-4

  • Empirical Formula (Hill Notation) C6H7F6N2P

  • Molecular Weight 252.10

  •  PubChem Substance ID 329751553

Purchase

Properties

InChI Key   INXUHWUWYCCCDE-UHFFFAOYSA-O

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?