EMAIL THIS PAGE TO A FRIEND
17747 Aldrich

Lutensol® FSA 10

Synonym: N-Oleylimino-bis(ethyl-polyethyleneglycol), Oleic amid-ethoxylate

Purchase

Properties

Related Categories Biochemicals and Reagents, Detergents, Detergents A to M, Non-Ionic Detergents
description   non-ionic
impurities   ≤1% water
density   1.02 g/mL at 20 °C

Description

Legal Information

Lutensol is a registered trademark of BASF SE

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?