• USA Home
  • 177563 - Dimethoxycarbenium hexafluorophosphate

EMAIL THIS PAGE TO A FRIEND
177563 Aldrich

Dimethoxycarbenium hexafluorophosphate

  • CAS Number 50318-32-6

  • Empirical Formula (Hill Notation) C3H7F6O2P

  • Molecular Weight 220.05

Purchase

Description

Application

A versatile alkylating agent.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?