• USA Home
  • 17756 - N1-Boc-3,3′-iminodipropylamine

EMAIL THIS PAGE TO A FRIEND
17756 Aldrich

N1-Boc-3,3′-iminodipropylamine

≥97.0% (NT)

Synonym: tert-Butyl [3-(3-aminopropylamino)propyl]carbamate

  • CAS Number 82409-04-9

  • Empirical Formula (Hill Notation) C11H25N3O2

  • Molecular Weight 231.34

  •  MDL number MFCD03788175

  •  PubChem Substance ID 24850370

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Linkers and Crosslinkers, Trifunctional Linkers
InChI Key   DZFPOUXCPWBUDA-UHFFFAOYSA-N
assay   ≥97.0% (NT)
refractive index   n20/D 1.474
density   1.001 g/mL at 20 °C(lit.)

Description

Other Notes

Building block for the synthesis of polyamines, e.g. polyamine toxins

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2734PSN1 3(8) / PGII
WGK Germany 
3
Flash Point(F) 
257 °F
Flash Point(C) 
125 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?