• USA Home
  • 18196 - (R)-4-Chloro-α-methylbenzylamine

EMAIL THIS PAGE TO A FRIEND
18196 Aldrich

(R)-4-Chloro-α-methylbenzylamine

≥95.0%

Synonym: (R)-1-(4-Chlorophenyl)ethylamine

  • CAS Number 27298-99-3

  • Empirical Formula (Hill Notation) C8H10ClN

  • Molecular Weight 155.62

  •  Beilstein Registry Number 2412317

  •  MDL number MFCD00671639

  •  PubChem Substance ID 57647722

Purchase

Properties

Related Categories Amines, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Chiral Resolution Reagents,
InChI Key   PINPOEWMCLFRRB-ZCFIWIBFSA-N
assay   ≥95.0%
optical activity   [α]/D +26.0±2.0°, c = 5 in ethanol
refractive index   n20/D 1.543(lit.)
density   1.110 g/mL at 20 °C(lit.)

Description

Analysis Note

Ion pair reagent for HPLC/MS

Packaging

5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN2810 - class 6.1 - PG 2 - EHS - Toxic, liquids, organic, n.o.s., HI: all
WGK Germany 
3
Flash Point(F) 
239 °F
Flash Point(C) 
115 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?