EMAIL THIS PAGE TO A FRIEND
182516 Aldrich

Poly(vinyl alcohol)

average Mw 106,000-110,000, powder, 100% hydrolyzed

Purchase

Properties

form   powder
mol wt   average Mw 106,000-110,000
transition temp   Tm 258 °C
  Tg 99 °C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?