• USA Home
  • 182990 - Poly(vinyl chloride-co-vinyl acetate-co-vinyl alcohol)

EMAIL THIS PAGE TO A FRIEND
182990 Aldrich

Poly(vinyl chloride-co-vinyl acetate-co-vinyl alcohol)

powder

Synonym: Vinyl chloride/vinyl acetate/vinyl alcohol terpolymer

Purchase

Properties

form   powder

Description

Warning

May contain vinyl chloride, an OSHA carcinogen. See 29 CFR part 1910

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?