• USA Home
  • 183881 - 2′-Hydroxy-4′,6′-dimethylacetophenone

EMAIL THIS PAGE TO A FRIEND
183881 Aldrich

2′-Hydroxy-4′,6′-dimethylacetophenone

97%

  • CAS Number 16108-50-2

  • Empirical Formula (Hill Notation) C10H12O2

  • Molecular Weight 164.20

  •  PubChem Substance ID 329751667

Purchase

Properties

InChI Key   AAPOVDIBCPRZPC-UHFFFAOYSA-N
assay   97%

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?