EMAIL THIS PAGE TO A FRIEND
184047 Aldrich

(S)-(+)-Leucinol

96%

Synonym: (S)-2-Amino-4-methyl-1-pentanol, 2-Amino-4-methyl-1-pentanol, L-Leucinol

Purchase

Properties

Related Categories Amino Acids & Derivatives, Amino Alcohols, Chemical Biology, Chemical Synthesis, Peptide Chemistry More...
InChI Key   VPSSPAXIFBTOHY-LURJTMIESA-N
assay   96%
optical activity   [α]20/D +4°, c = 9 in ethanol
optical purity   ee: 99% (GLC)
refractive index   n20/D 1.4511(lit.)
bp   198-200 °C/768 mmHg(lit.)
density   0.917 g/mL at 25 °C(lit.)

Description

Packaging

5, 25 g in glass bottle

Application

Starting material for the synthesis of aminopeptidase N and phospholipase A2 inhibitors.

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
195.8 °F
Flash Point(C) 
91 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?