• USA Home
  • 184160 - 4-Chloro-3-nitrobenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
184160 Aldrich

4-Chloro-3-nitrobenzyl alcohol

98%

Purchase

Properties

Related Categories Alcohols, Building Blocks, C7 to C8, Chemical Synthesis, Organic Building Blocks,
InChI Key   QLLRQJDSYJIXTN-UHFFFAOYSA-N
assay   98%
mp   62-64 °C(lit.)

Description

Packaging

5 g in glass bottle

Application

4-Chloro-3-nitrobenzyl alcohol was used in the preparation of 4-chloro-3-nitrobenzyl hepta-O-acetyl-β-maltoside and 3-iodothyronamine.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?