• USA Home
  • 184322 - 4-Methoxyphenyl methyl sulfone

EMAIL THIS PAGE TO A FRIEND
184322 Aldrich

4-Methoxyphenyl methyl sulfone

95%

  • CAS Number 3517-90-6

  • Empirical Formula (Hill Notation) C8H10O3S

  • Molecular Weight 186.23

  •  PubChem Substance ID 329751669

Purchase

Properties

InChI Key   KAZUCVUGWMQGMC-UHFFFAOYSA-N
assay   95%

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?