EMAIL THIS PAGE TO A FRIEND
185604 Aldrich

Naphthalene

98%

  • CAS Number 91-20-3

  • Empirical Formula (Hill Notation) C10H8

  • Molecular Weight 128.17

  •  Beilstein Registry Number 1421310

  •  EC Number 202-049-5

  •  MDL number MFCD00001742

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Arenes, Building Blocks, Chemical Synthesis, Organic Building Blocks
vapor density   4.4 (vs air)
vapor pressure   0.03 mmHg ( 25 °C)
InChI Key   UFWIBTONFRDIAS-UHFFFAOYSA-N
assay   98%
autoignition temp.   978 °F
expl. lim.   5.9 %
bp   218 °C(lit.)
mp   80-82 °C(lit.)

Description

Other Notes

This product has been replaced by 147141-ALDRICH | Naphthalene 99%

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN1334 - class 4.1 - PG 3 - Naphthalene, crude
WGK Germany 
3
RTECS 
QJ0525000
Flash Point(F) 
176 °F
Flash Point(C) 
80 °C
Protocols & Articles

Related Content

Carcinogen Selector - Reproductive System - Lung

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Reproductive System - Lung
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Reproductive System - Nasal Cavity

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Reproductive System - Nasal Cavity
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Respiratory System

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Respiratory System
Keywords: Cancer, Carcinogens, Respiratory

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

147141 Naphthalene, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?