• USA Home
  • 187674 - 1-Cyclohexylcyclohexanol

EMAIL THIS PAGE TO A FRIEND
187674 Aldrich

1-Cyclohexylcyclohexanol

97%

  • CAS Number 2903-12-0

  • Empirical Formula (Hill Notation) C12H22O

  • Molecular Weight 182.30

  •  PubChem Substance ID 329751741

Purchase

Properties

InChI Key   PVSNMDAHWMHSBD-UHFFFAOYSA-N
assay   97%

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?