• USA Home
  • 189057 - 4-Benzyloxy-3-methoxybenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
189057 Aldrich

4-Benzyloxy-3-methoxybenzyl alcohol

97%

Purchase

Properties

Related Categories Alcohols, Building Blocks, C11 to C30+, Chemical Synthesis, Organic Building Blocks,
InChI Key   PDBXFVPMVYQICB-UHFFFAOYSA-N
assay   97%
mp   71-73 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?