EMAIL THIS PAGE TO A FRIEND
189375 Aldrich

Poly(vinyl alcohol)

average Mw 3,000, crystalline, 75% hydrolyzed

Purchase

Properties

form   crystalline
mol wt   average Mw 3,000

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?