• USA Home
  • 190330 - 1,2,3,6,7,8,11,12-Octahydrobenzo[e]pyren-9(10H)-one

EMAIL THIS PAGE TO A FRIEND
190330 Aldrich

1,2,3,6,7,8,11,12-Octahydrobenzo[e]pyren-9(10H)-one

95%

Purchase

Properties

Related Categories Building Blocks, C15 to C38, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   UIJOUFUKJPLQEK-UHFFFAOYSA-N
assay   95%
mp   146-148 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?