• USA Home
  • 190489 - 2,4,5-Trimethoxybenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
190489 Aldrich

2,4,5-Trimethoxybenzyl alcohol

97%

Purchase

Properties

Related Categories Alcohols, Building Blocks, C9 to C10, Chemical Synthesis, Organic Building Blocks,
InChI Key   XJNCXXBIDXLPMG-UHFFFAOYSA-N
assay   97%
mp   70-72 °C(lit.)

Description

Packaging

5 g in glass bottle

General description

Oxidation of 2,4,5-trimethoxybenzyl alcohol in the presence of laccase yields a dimeric compound.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?