• USA Home
  • 19159 - (S)-3-Butene-1,2-diol

EMAIL THIS PAGE TO A FRIEND
19159 Aldrich

(S)-3-Butene-1,2-diol

≥97.0% (GC)

  • CAS Number 62214-39-5

  • Empirical Formula (Hill Notation) C4H8O2

  • Molecular Weight 88.11

  •  Beilstein Registry Number 4241981

  •  MDL number MFCD00145261

  •  PubChem Substance ID 24851495

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks, Polyols More...
InChI Key   ITMIAZBRRZANGB-BYPYZUCNSA-N
assay   ≥97.0% (GC)
refractive index   n20/D 1.461
density   1.053 g/mL at 20 °C(lit.)
storage temp.   2-8°C

Description

Packaging

100 mg in glass bottle

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
228.2 °F
Flash Point(C) 
109 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?