• USA Home
  • 19163 - (S)-3-Butene-1,2-diol-1-(p-toluenesulfonate)

EMAIL THIS PAGE TO A FRIEND
19163 Aldrich

(S)-3-Butene-1,2-diol-1-(p-toluenesulfonate)

≥99.0% (HPLC)

Synonym: (S)-2-Hydroxy-3-butenyl tosylate, (S)-3-Butene-1,2-diol 1-tosylate, (S)-1-Tosyloxy-3-buten-1-ol

  • CAS Number 133095-74-6

  • Empirical Formula (Hill Notation) C11H14O4S

  • Molecular Weight 242.29

  •  Beilstein Registry Number 4254384

  •  MDL number MFCD00145259

  •  PubChem Substance ID 24851498

Purchase

Properties

Related Categories Alcohols, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   YQSCDBZHHLIPOI-JTQLQIEISA-N
assay   ≥99.0% (HPLC)
optical activity   [α]20/D −7.7±0.5°, c = 1% in methanol
storage temp.   2-8°C

Description

Packaging

1 g in glass bottle

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?