• USA Home
  • 192775 - Diethylaluminum iodide solution

EMAIL THIS PAGE TO A FRIEND
192775 Aldrich

Diethylaluminum iodide solution

25 wt. % in toluene

Purchase

Properties

concentration   25 wt. % in toluene

Description

Packaging

Packaged under nitrogen in poly-coated Sure/Seal bottles

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 3394 4.2/PG 1
Flash Point(F) 
-1 °F
Flash Point(C) 
-18 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?