• USA Home
  • 194174 - 7-Aminodesacetoxycephalosporanic acid

EMAIL THIS PAGE TO A FRIEND
194174 Aldrich

7-Aminodesacetoxycephalosporanic acid

98%

Synonym: 7-ADCA

  • CAS Number 22252-43-3

  • Empirical Formula (Hill Notation) C8H10N2O3S

  • Molecular Weight 214.24

  •  MDL number MFCD09750368

Purchase

Properties

InChI Key   NVIAYEIXYQCDAN-CLZZGJSISA-N
assay   98%

Description

Application

Potent inhibitor of bacterial cell-wall enzyme.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?