-
Product Directory
Custom Product
-
Services Offered
Custom Capabilities
-
Synonym: Pentabromophenyl ether
CAS Number 1163-19-5
Linear Formula C6Br5OC6Br5
Molecular Weight 959.17
Beilstein Registry Number 2188438
EC Number 214-604-9
MDL number MFCD00000059
PubChem Substance ID 24851652
| Related Categories | Building Blocks, Chemical Synthesis, Ethers, Organic Building Blocks, Oxygen Compounds More... |
| assay | 98% |
| mp | >300 °C(lit.) |
100 g in poly btl
Customers Also Purchased
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?