• USA Home
  • 195006 - α,α,α′,α′-Tetrabromo-m-xylene

EMAIL THIS PAGE TO A FRIEND
195006 Aldrich

α,α,α′,α′-Tetrabromo-m-xylene

Purchase

Properties

Related Categories Aryl, Building Blocks, C8, Chemical Synthesis, Halogenated Hydrocarbons,
InChI Key   ZMCUKNMLHBAGMS-UHFFFAOYSA-N
mp   105-108 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3448 6.1 / PGII
WGK Germany 
3
RTECS 
ZE5150000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?