• USA Home
  • 196584 - 2,3,4,5,6,2′,3′,4′,5′,6′-Decafluorobenzhydrol

EMAIL THIS PAGE TO A FRIEND
196584 Aldrich

2,3,4,5,6,2′,3′,4′,5′,6′-Decafluorobenzhydrol

≥97%

Synonym: 2,3,4,5,6,2′,3′,4′,5′,6′-Decafluorodiphenylcarbinol, Bis(pentafluorophenyl)methanol

Purchase

Properties

Related Categories Alcohols, Building Blocks, C11 to C30+, Chemical Synthesis, Organic Building Blocks,
InChI Key   WRLLBTKDSCJOBL-UHFFFAOYSA-N
assay   ≥97%
mp   77-80 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?