• USA Home
  • 197750 - (±)-10-Camphorsulfonic acid monohydrate

EMAIL THIS PAGE TO A FRIEND
197750 Aldrich

(±)-10-Camphorsulfonic acid monohydrate

98%

Purchase

Properties

assay   98%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?