• USA Home
  • 20024 - N-Butyl-2,2′-imino-di(8-quinolinol)

EMAIL THIS PAGE TO A FRIEND
20024 Aldrich

N-Butyl-2,2′-imino-di(8-quinolinol)

≥98.0%

Synonym: N-Butyl-2,2′-imino-bis(8-hydroxyquinoline)

  • CAS Number 82361-90-8

  • Empirical Formula (Hill Notation) C22H21N3O2

  • Molecular Weight 359.42

  •  MDL number MFCD00168944

  •  PubChem Substance ID 24851354

Purchase

Properties

Related Categories Building Blocks, C11 to C30, Chemical Synthesis, Heterocyclic Building Blocks, Quinolines More...
InChI Key   AWKZSNHJRJMXCN-UHFFFAOYSA-N
assay   ≥98.0%
mp   93-96 °C

Description

Packaging

500 mg in glass bottle

Application

N-Butyl-2,2′-imino-di(8-quinolinol) may be used in chemical synthesis studies.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?