• USA Home
  • 200255 - Poly(ethylene terephthalate)

EMAIL THIS PAGE TO A FRIEND
200255 Aldrich

Poly(ethylene terephthalate)

pellets, average Mv ~18,000

Purchase

Properties

form   pellets
mol wt   average Mv ~18,000
intrinsic viscosity   0.59(lit.)
transition temp   Tg 81 °C
density   1.375 g/mL at 25 °C

Description

Application

Used in electrical parts including relay bases and lamp sockets, pump housings, gears, sprockets, chair arms, casters and furniture components.

Features and Benefits

Balance of strength, stiffness and toughness, excellent electrical properties, surface appearance and chemical resistance.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?