• USA Home
  • 200301 - Poly(vinyl chloride-co-vinyl acetate-co-maleic acid)

EMAIL THIS PAGE TO A FRIEND
200301 Aldrich

Poly(vinyl chloride-co-vinyl acetate-co-maleic acid)

average Mw ~70,000 by GPC, powder

  • CAS Number 9005-09-8

  • Linear Formula [CH2CH(Cl)]x[CH2CH(O2CCH3)]y[CH(CO2H)CH(CO2H)]Z

  •  MDL number MFCD00134024

Purchase

Properties

form   powder
mol wt   average Mw ~70,000 by GPC
density   1.35 g/mL at 25 °C(lit.)

Description

Warning

May contain vinyl chloride, an OSHA carcinogen. See 29 CFR part 1910

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?