• USA Home
  • 200344 - Poly(isobutyl vinyl ether)

EMAIL THIS PAGE TO A FRIEND
200344 Aldrich

Poly(isobutyl vinyl ether)

average Mw ~600,000, average Mn ~170,000

Purchase

Properties

form   chunks
mol wt   average Mn ~170,000
  average Mw ~600,000

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?