• USA Home
  • 200352 - Poly[methylene(polyphenyl isocyanate)]

EMAIL THIS PAGE TO A FRIEND
200352 Aldrich

Poly[methylene(polyphenyl isocyanate)]

average Mw 360

Synonym: PAPI® 135

Purchase

Properties

vapor pressure   <10-5 mmHg ( 25 °C)
activity   31.5% isocyanate
mol wt   average Mw 360
viscosity   185 cP(25 °C)(lit.)

Description

Legal Information

PAPI is a registered trademark of Upjohn Co.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?