Synonym: Basic Blue 16
CAS Number 4569-88-4
Empirical Formula (Hill Notation) C30H24ClN5O
Molecular Weight 506.00
Colour Index Number 12210
EC Number 224-951-8
MDL number MFCD00011924
PubChem Substance ID 24851979
| Related Categories | Materials Science, Organic and Printed Electronics, Photochromic and Thermochromic Dyes, Photonic and Optical Materials |
| composition | Dye content, 70% |
| UV absorption | λmax 589 nm |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?