• USA Home
  • 20212 - 4-(tert-Butoxymethyl)benzoic acid

EMAIL THIS PAGE TO A FRIEND
20212 Aldrich

4-(tert-Butoxymethyl)benzoic acid

≥98.0% (T)

  • CAS Number 34224-31-2

  • Empirical Formula (Hill Notation) C12H16O3

  • Molecular Weight 208.25

  •  Beilstein Registry Number 2719166

  •  MDL number MFCD01862971

  •  PubChem Substance ID 24852039

Purchase

Properties

Related Categories Bifunctional Linkers, Chemical Biology, Chemical Synthesis, Linkers and Crosslinkers
InChI Key   HXNWRKOKHLXUQN-UHFFFAOYSA-N
assay   ≥98.0% (T)
mp   147-151 °C

Description

Other Notes

Linker for the solid-phase synthesis of peptidomimetics

Packaging

2.5 g in poly bottle

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?