MDL number MFCD00143923
PubChem Substance ID 24852421
Popular Documents: RAMAN FTIR (PDF)
| Related Categories | C, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes |
| UV absorption | λmax 547 nm |
5, 25 g in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?