• USA Home
  • 207187 - Holmium atomic absorption standard solution

EMAIL THIS PAGE TO A FRIEND
207187 Aldrich

Holmium atomic absorption standard solution

1,000 μg/mL Ho in 1 wt. % HNO3 (Exact Ho concentration on label)

Purchase

Properties

concentration   1,000 μg/mL Ho in 1 wt. % HNO3 (Exact Ho concentration on label)
application(s)   ICP/AAS: suitable
format   single component solution

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 3264 8/PG 3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?