• USA Home
  • 207667 - Zinc atomic absorption standard solution

EMAIL THIS PAGE TO A FRIEND
207667 Aldrich

Zinc atomic absorption standard solution

1,000 μg/mL Zn+2 in 1 wt. % HCl (Exact Zn concentration on label)

Purchase

Properties

Related Categories AA Standard Solutions, AAS, Additional Standards, Analytical Standards, Analytical/Chromatography,
concentration   1,000 μg/mL Zn+2 in 1 wt. % HCl (Exact Zn concentration on label)
application(s)   ICP/AAS: suitable
density   1.01 g/mL at 25 °C
format   single component solution

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1789 8 / PGIII
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

18827 Zinc Standard for AAS, TraceCERT®, 1000 mg/L Zn in nitric acid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?