EMAIL THIS PAGE TO A FRIEND
207721 Aldrich

Guinea Green B

Dye content 50 %

Synonym: Acid Green 3

  • CAS Number 4680-78-8

  • Empirical Formula (Hill Notation) C37H35N2NaO6S2

  • Molecular Weight 690.80

  •  Colour Index Number 42085

  •  EC Number 225-132-8

  •  MDL number MFCD00012122

  •  PubChem Substance ID 24852468

  • Popular Documents:  FTIR (PDF)  

Purchase

Properties

Related Categories Cell Biology, G-H-I, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes More...
InChI Key   XKTMIJODWOEBKO-UHFFFAOYSA-M
composition   Dye content, 50%
mp   255 °C (dec.)(lit.)
absorption   λmax 618 nm

Description

Packaging

25 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Carcinogen Selector - Digestive System - Liver

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Digestive System - Liver
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System - Liver Endocrine

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Endocrine System - Liver Endocrine
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Endocrine System
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Hematopoietic System

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Hematopoietic System
Keywords: Cancer, Carcinogens, Respiratory

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?