• USA Home
  • 209570 - 2,3-Dimethyl-4-nitroanisole

EMAIL THIS PAGE TO A FRIEND
209570 Aldrich

2,3-Dimethyl-4-nitroanisole

95%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitro Compounds, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   DUBFMQLUMHKYOX-UHFFFAOYSA-N
assay   95%
mp   70-73 °C(lit.)

Description

Packaging

10 g in poly bottle

Application

2,3-Dimethyl-4-nitroanisole was used:
• as starting material in the synthesis of conformationally restricted derivatives of lavendustin A
• in the synthesis of 1,4-piperazine-2,5-diones

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?