• USA Home
  • 209902 - N-Ethyl-2,3-xylidine

EMAIL THIS PAGE TO A FRIEND
209902 Aldrich

N-Ethyl-2,3-xylidine

99%

  • CAS Number 41115-23-5

  • Empirical Formula (Hill Notation) C10H15N

  • Molecular Weight 149.23

  •  PubChem Substance ID 329752067

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   IILPUHXLVPEPAV-UHFFFAOYSA-N
assay   99%

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?