• USA Home
  • 211206 - N,N-Bis(2-hydroxyethyl)-1,4-phenylenediamine sulfate salt monohydrate

EMAIL THIS PAGE TO A FRIEND
211206 Aldrich

N,N-Bis(2-hydroxyethyl)-1,4-phenylenediamine sulfate salt monohydrate

97%

  • CAS Number 57524-61-5

  • Empirical Formula (Hill Notation) C10H16N2O2 · H2O · H2SO4

  • Molecular Weight 312.34

  •  PubChem Substance ID 329752083

Purchase

Properties

InChI Key   ZWZDWBQKZSSBNM-UHFFFAOYSA-N
assay   97%

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?