EMAIL THIS PAGE TO A FRIEND
211850 Aldrich

Sulfamethazine

99%

Synonym: N1-(4,6-Dimethyl-2-pyrimidinyl)sulfanilamide

Purchase

Properties

assay   99%

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
2
RTECS 
WO9275000
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?