EMAIL THIS PAGE TO A FRIEND
215775 Aldrich

Nickel

spheres, 6-16 mm, ≥99.95% trace metals basis

Purchase

Properties

Related Categories 28: Ni, Magnetic Materials, Magnetic Precursors, Materials Science, Metal and Ceramic Science,
InChI Key   PXHVJJICTQNCMI-UHFFFAOYSA-N
assay   ≥99.95% trace metals basis
form   spheres
resistivity   6.97 μΩ-cm, 20°C
particle size   6-16 mm
bp   2732 °C(lit.)
mp   1453 °C(lit.)
density   8.9 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
QR5950000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?