• USA Home
  • 217301 - Sodium iodide dihydrate

EMAIL THIS PAGE TO A FRIEND
217301 Aldrich

Sodium iodide dihydrate

≥99%

Purchase

Properties

Related Categories Chemical Synthesis, Essential Chemicals, Inorganic Salts, Materials Science, Metal and Ceramic Science,
vapor density   >1 (vs air)
InChI Key   VEEIYHUOLCUPSA-UHFFFAOYSA-M
assay   ≥99%
form   crystalline
density   2.448 g/mL at 25 °C(lit.)

Description

Packaging

100 g in glass bottle

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?