• USA Home
  • 218073 - Tilorone analog R10,874DA

EMAIL THIS PAGE TO A FRIEND
218073 Aldrich

Tilorone analog R10,874DA

Synonym: 3,6-Bis[2-(Dimethylamino)ethoxy]-9H-xanthen-9-one dihydrochloride, 3,6-Bis[2-(dimethylamino)ethoxy]-9H-xanthen-9-one dihydrochloride

Purchase

Properties

InChI Key   TYTSHTWUNBMVBR-UHFFFAOYSA-N

Description

Application

These compounds are a group of bis-basic-substituted polycyclic aromatic compounds structurally and functionally related to Tilorone. They offer research opportunities in the areas of interferon induction, immunoregulation, molecular genetics, viral/cancer chemotherapy, etc. A bibliography of over 300 references on Tilorone and analogs is available on request.

Biochem/physiol Actions

Tilorone (2,7-bis[2-(diethylamino)ethoxy]-9H-fluoren-9-one) was the first low-m.w. synthetic interferon inducer.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?