• USA Home
  • 221120 - N-(4-Aminobenzoyl)-L-glutamic acid diethyl ester

EMAIL THIS PAGE TO A FRIEND
221120 Aldrich

N-(4-Aminobenzoyl)-L-glutamic acid diethyl ester

97%

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Glutamic Acid, Peptide Chemistry More...
InChI Key   RJXFBLRRPYBPTM-ZDUSSCGKSA-N
assay   97%
optical activity   [α]21/D +17.9°, c = 5 in chloroform
mp   139-142 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?