EMAIL THIS PAGE TO A FRIEND
222844 Aldrich

DL-Norvaline

99%

Synonym: (±)-2-Aminopentanoic acid

  • CAS Number 760-78-1

  • Linear Formula CH3CH2CH2CH(NH2)COOH

  • Molecular Weight 117.15

  •  Beilstein Registry Number 1721163

  •  EC Number 212-082-7

  •  MDL number MFCD00064420

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   SNDPXSYFESPGGJ-UHFFFAOYSA-N
assay   99%

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

N7502 DL-Norvaline

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?