EMAIL THIS PAGE TO A FRIEND
223093 Aldrich

Sarcosine anhydride

99%

Synonym: 1,4-Dimethyl-2,5-piperazinedione

  • CAS Number 5076-82-4

  • Empirical Formula (Hill Notation) C6H10N2O2

  • Molecular Weight 142.16

  •  Beilstein Registry Number 123513

  •  EC Number 225-788-5

  •  MDL number MFCD00006519

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   PKDGRAULLDDTRN-UHFFFAOYSA-N
assay   99%
mp   144-147 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?